Butein ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 2',3,4,4'-Tetrahydroxychalcone, (E)-1-(2,4-Dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)-2-propen-1-one

350-246-M010
89149-324EA 301.93 USD
89149-324
Butein ≥98% (by HPLC)
Butein

Tyrosine kinase inhibitor


Plant polyphenol. Specific tyrosine kinase inhibitor. Potently inhibits the tyrosine kinase activity of the EGF receptor and p60c-src. Potent antioxidant and anti-inflammatory agent. Inhibits glutathione reductase and rat liver glutathione S-transferase. Activator of human deacetylase SIRT1. Inhibits aromatase, showing chemopreventive properties. Directly inhibits IKK.

Formula: C₁₅H₁₂O₅
MW: 272.26 g/mol
MDL Number: MFCD00017300
CAS Number: 487-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8067caee4ceeb7be216bd3b7579b7999fbe101da&vid=265e29670a7221c76748d24b49b10c7953dd0f26&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR