Chlortetracycline hydrochloride ≥80%

Supplier: Enzo Life Sciences

Synonyms: Chlortetracycline HCI, Aureomycin, 7-Chlorotetracycline hydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride

350-238-G005
89149-310EA 70.85 USD
89149-310
Chlortetracycline hydrochloride ≥80%
Chlortetracycline hydrochloride

Protein synthesis inhibitor


Antibiotic. Inhibits protein synthesis in prokaryotes and eukaryotes. Inhibits binding of aminoacyl-tRNA to ribosomes. Used as a fluorescent Ca2+ probe.


Isolated from Streptomyces aureofaciens.

Formula: C₂₂H₂₃ClN₂O₈·HCl
MW: 515.34 g/mol
MDL Number: MFCD26142699
CAS Number: 64-72-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6636724c3ac146febc75100e810fbb8d81faa85a&vid=583315bc1d86b7f6105235b69354d20cc6ef1fd1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR