Rosiglitazone ≥97% (by HPLC)

Supplier: Enzo Life Sciences

350-125-M025 350-125-M100
89149-120EA 170.04 USD
89149-120 89149-122
Rosiglitazone ≥97% (by HPLC)
Rosiglitazone

Insulin sensitizing agent.


  • Formulation: White to off-white solid.


Peroxisome proliferator-activated receptor γ (PPAR-γ) agonist. Hepatotoxic. Antidiabetic.


Soluble in DMSO (>25 mg/mL), 100% ethanol or methanol.

Formula: C₁₈H₁₉N₃O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00871760
CAS Number: 122320-73-4
Merck Index: 14,08265
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb8b795042af6f8cb409adc6367d9a353cadb008&vid=1c114a0f89ab412a2cf78f3ec4c3bfc809b296a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥97% by HPLC


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR