Dihydrocapsaicin ≥85% (by HPLC)

Supplier: Enzo Life Sciences

350-052-M050
89148-982EA 175.49 USD
89148-982
Dihydrocapsaicin ≥85% (by HPLC)
Dihydrocapsaicin

Dihydro-analog and congener of capsaicin (Prod. No. BML-EI125) in chili peppers (Capsicum). Like capsaicin it is irritant. Dihydrocapsaicin accounts for about 22% of the total capsaicinoids mixture and has about the same pungency as capsaicin. Antioxidant. Reduces oxidation of serum lipids. Mutagenic.


  • Formulation: White to off-white solid
  • Isolated from Capsicum fruit


Isolated from Capsicum fruit.

Formula: C₁₈H₂₉NO₃
MDL Number: MFCD00057794
CAS Number: 19408-84-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbd131628d1300116bd5ef0a0667b2fdcca1a3d9&vid=26a82e63620e888b79f6c4414d861e181e8d65b6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR