PQ401 ≥98% (by HPLC and UV method)

Supplier: Enzo Life Sciences

Synonyms: 1-(5-Chloro-2-methoxyphenyl)-3-(2-methylquinolin-4-yl)urea, N-(5-Chloro-2-methoxyphenyl)-N’-(2-methyl-4-quinolinyl)urea, IGF-IR Inhibitor

270-459-M005
89147-962EA 174.4 USD
89147-962
PQ401 ≥98% (by HPLC and UV method)
PQ401

Potent inhibitor of insulin-like growth factor I receptor (IGF-IR) signalling and breast cancer cell growth in culture and in vivo.


  • IGF-1 receptor inhibitor


Off-white solid.


Soluble in DMSO (10mg/ml) or methanol (7mg/ml).

Formula: C₁₈H₁₆ClN₃O₂
MW: 341.8 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00160558
CAS Number: 196868-63-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6367eaaec2387d440d05c8ba690df523cb944f5a&vid=12688e7939b3038404e99d76ae1dd9e9d1bbf8da&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR