SANT-2 ≥95%

Supplier: Enzo Life Sciences

Synonyms: N-(3-(1H-Benzo[d]imidazol-2-yl)-4-chlorophenyl)-3,4,5-triethoxybenzamide

270-436-M001
89147-898EA 292.12 USD
89147-898
SANT-2 ≥95%
SANT-2

Smoothened inhibitor


Cell permeable antagonist of the Smoothened (Smo) protein function. Inhibitor of SAG (Prod. No. ALX-270-426) activity. Chemical tool to study the Hedgehog (Hh) signalling pathway and to explore therapeutic uses for inhibitors of Smo.

Formula: C₂₆H₂₆ClN₃O₄
MW: 480 g/mol
MDL Number: MFCD01827636
CAS Number: 329196-48-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebf2338070ea75e08a6f393dbf0e3e2ee9a89c74&vid=795edcb753272be20135d374e088516bd295689a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR