Salubrinal ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
(E)-N-(2,2,2-Trichloro-1-(3-(quinolin-8-yl)thioureido)ethyl)cinnamamide
270-428-M001
270-428-M005
89147-876EA
50.14
USD
89147-876
89147-878
Salubrinal ≥98% (by HPLC)
Salubrinal
Cell permeable and selective inhibitor of the phosphatase complexes that dephosphorylate eukaryotic translation initiation factor 2 subunit alpha (eIF-2alpha). Protects ER stress-induced apoptosis (EC50~15µM in PC12 cells stimulated with 750ng/ml of tunicamycin (Prod. No. BML-CC104)).
Formula:
C₂₁H₁₇Cl₃N₄OS MW: 479.81 g/mol |
MDL Number:
MFCD00548612 CAS Number: 405060-95-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...