Anacardic acid ≥95% (by 1H-NMR)

Supplier: Enzo Life Sciences

Synonyms: 2-Hydroxy-6-pentadecylbenzoic acid

270-381-M005 270-381-M025
89147-748EA 75.21 USD
89147-748 89147-750
Anacardic acid ≥95% (by 1H-NMR)
Anacardic acid

Cell permeable salicylic acid analog that acts as a potent, non-competitive inhibitor of p300 and PCAF (p300/CBP-associated factor) histone acetyltransferase (HAT) activities (IC50~8.5µM and ~5µM, respectively). Displays a variety of other biological activities, such as antibacterial, antimicrobial, prostaglandin synthase inhibition, tyrosinase, and lipoxygenase inhibition.Inhibits SUMOylation of RanGAP1-C2 with an IC50 = 2.2µM


Synthetic.

Formula: C₂₂H₃₆O₃
MDL Number: MFCD07368254
CAS Number: 16611-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f72901076c3398eb02ec68368248eee4e206c9d&vid=841e72401428e40ea7af1d9e00c48a134e5ec126&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR