2,2-Bis(4-glycidyloxyphenyl)propane ≥85%

Supplier: Enzo Life Sciences

Synonyms: Bisphenol A Diglycidyl Ether, 2,2-Bis(4-hydroxyphenyl)propane Diglycidyl Ether, DER 332, BADGE, D.E.R. 332 epoxy resin

270-353-G025
89147-684EA 54.5 USD
89147-684
2,2-Bis(4-glycidyloxyphenyl)propane ≥85%
2,2-Bis(4-glycidyloxyphenyl)propane

PPARgamma inhibitor


Antagonist of PPARgamma. Binds to PPARgamma with an apparent Kd of 100µM.

Formula: C₂₁H₂₄O₄
MW: 340.42 g/mol
MDL Number: MFCD00080480
CAS Number: 1675-54-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fa2fc0c60089d64bdc13cb003e848fe876e2b1f&vid=0d7be87c42ccf0950761d61ee4acc01ca335923d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR