(-)-Epigallocatechin gallate (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate, (2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate, EGCG

270-263-M010 270-263-M050
89147-516EA 49.05 USD
89147-516 89147-518
(-)-Epigallocatechin gallate (by HPLC)
(-)-Epigallocatechin gallate

Inhibitor of NOS, telomerase and Dnmt


Antitumor reagent. Antioxidant. Protects cells from lipid peroxidation and DNA damage induced by reactive free radicals. Inhibits inducible nitric oxide synthase (iNOS; NOS II). Chemopreventive anticancer agent. Induces apoptosis in human cancer cell lines. Inhibits MAP kinase mediated signalling pathways. Inhibits angiogenesis. Inhibits telomerase and DNA methyltransferase. Anti-inflammatory agent.


Isolated from green tea.

Formula: C₂₂H₁₈O₁₁
MW: 458.38 g/mol
MDL Number: MFCD00075940
CAS Number: 989-51-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e75e239727741f9ac8c18b25c38811e39764e73&vid=36f176b4256bbb9f93136620c928fa622c1c617b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR