Chlorpromazine hydrochloride ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 2-Chloro-10-[3-(dimethylamino)-1-propyl]phenothiazine Hydrochloride

270-171-G005
89147-318EA 75.21 USD
89147-318
Chlorpromazine hydrochloride ≥98% (by HPLC)
Chlorpromazine hydrochloride

Antipsychotic working on a variety of receptors in the CNS


Inhibits calmodulin stimulation of cyclic nucleotide phosphodiesterase. Dopamine antagonist. D2 antagonist. H1 antagonist. Inhibits TNF-alpha production. Potent PLA2 inhibitor. Inhibits nitric oxide synthase (NOS) in mouse brain and prevents lipopolysaccharide induction of NOS in mouse lung. Antipsychotic.

Formula: C₁₇H₁₉ClN₂S
MW: 355.32 g/mol
MDL Number: MFCD00012654
CAS Number: 69-09-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b22ff716bb4b4e0998de7e4b6bdf5f9f85f8cc83&vid=2c4bd36ff17b86d345f28b6d74a9f363e9f004f7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR