Anti-APRIL Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-950-C100
89146-348EA 488.32 USD
89146-348
Anti-APRIL Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes membrane-bound form of human APRIL. Does not recognize the soluble form of human APRIL.

Type: Primary
Antigen: APRIL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR