Anti-ACTN4 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-356-C050
89145-522EA 963.56 USD
89145-522
Anti-ACTN4 Rabbit Polyclonal Antibody
Antibodies
α-Actinin-4 is an actin-bundling protein of ~100kDa that is associated with cell motility, endocytosis, and cancer invasion. The α-actinin family comprises two non-muscle isoforms (α-actinin-1 and -4) and two skeletal muscle isoforms (α-actinin-2 and -3), with α-actinin-2 being also expressed in cardiac muscle. While α-actinin-4 is almost ubiquitously expressed, particularly high concentrations are found in glomeruli. On the subcellular level it is associated with actin stress fibers, but in certain cells it also localizes to the nucleus. Mutations in the α-actinin-4 gene cause an autosomal-dominant form of familial focal segmental glomerulosclerosis (FSGS), which is thought to result from a defect in glomerular podocyte function. A point mutation in the α-actinin-4 gene was found to generate an antigenic peptide that is recognized by autologous cytolytic T lymphocytes (CTL) on a human lung carcinoma. α-actinin-4 interacts with a variety of proteins, including the ring finger protein BERP, the PDZ-LIM protein CLP-36, the hemidesmosomal and cell-cell contact protein BP180, and the tight junction protein MAGI-1. Moreover, α-actinin-4 forms a ternary complex with Ca2+/ Calmodulin-dependent protein kinase II and Densin-180, a protein of postsynaptic densities in CNS neurons. Ca2+-dependent association of α-actinin-4 with E3KARP is required for Ca2+-dependent inhibition of the Na+/ H+exchanger 3 (NHE3).

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, pig, and Chinese hamster actinin-4. Does not cross-react with actinin-1, -2 or -3.

Type: Primary
Antigen: ACTN4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR