PGE2 CLIA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-910-001
89141-748EA 588.6 USD
89141-748
PGE2 CLIA Kit, Enzo Life Sciences
Assays Immunological Assays
For the quantitative determination of PGE2 in culture supernatants, serum, saliva, urine, and whole blood from any species.
The PGE2 CLIA kit is a sensitive chemiluminescent competitive enzyme immunoassay kit for the quantitative determination of PGE2 in biological fluids from any species.
Ordering information: Contains White GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, CLIA Substrate.