Hydrogen Peroxide Fluorometric Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-907-028
89141-682EA 536.28 USD
89141-682
Hydrogen Peroxide Fluorometric Detection Kit, Enzo Life Sciences
Assays Cellular Assays
The Hydrogen Peroxide fluorometric detection kit provides fast results in just 10 minutes for the measurement of either hydrogen peroxide or peroxidase activity in a convenient one-step homogeneous assay.
  • Flexible in measure hydrogen peroxide or peroxidase activity
  • Higher throughput analysis
  • Fast results
For the measurement of hydrogen peroxide in cell lysates and culture supernatants from any species.
Delivery information: Reaction buffer concentrate, Detection Reagent, Hydrogen peroxide, Horseradish peroxidase