Catalase Fluorometric Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-907-027
89141-680EA 527.56 USD
89141-680
Catalase Fluorometric Detection Kit, Enzo Life Sciences
Assays Cellular Assays
The Catalase fluorometric detection kit provides a higher throughput sensitive method for measuring catalase activity in just one hour
  • Easily adaptable to HTS applications
  • Sensitive fluorescent detection system
For the measurement of catalase in cell lysates from any species.
This is a homogeneous mix-and-read assay that enables you to monitor multiple time points for a kinetic read. Positive control is provided.
Delivery information: Reaction buffer concentrate, Detection Reagent, Horseradish peroxidase concentrate, Hydrogen peroxide, Catalase enzyme