PGE2 ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-901-001
89141-268EA 2066.64 USD
89141-268
PGE2 ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive PGE2 ELISA kit for inflammation and eicosanoid research.
For the quantitative determination of PGE2 in culture supernatants, serum, saliva, urine, and whole blood from any species. Cited sample types include cell lysate, cerebral spinal fluid, dialysate, gingival crevicular fluid, peritoneal exudate fluid and peritoneal exudate cell supernatant, plasma, and tissue.
The PGE2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 2. Commercially available since 1992, this kit is widely cited in peer-reviewed publications. The non-radioactive ready-to-use liquid color-coded reagents reduce errors.
Ordering information: Contains GxM Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-052 (1x96 wells).
Caution: Store all components at +4 °C, except standard and conjugate at –20 °C.