Progesterone ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-011
89141-064EA 413.11 USD
89141-064
Progesterone ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of progesterone in culture supernatants, serum, and saliva from any species
The Progesterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in <3 hours. Absorbance is read at 405 nm. This non-radioactive enzyme immunoassay is safe and reliable with ready-to-use liquid color-coded reagents to reduce errors.
Cited sample type includes plasma.
Ordering information: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent. Alternative size available: 89141-280 (5x96 wells).