BRAND® Imhoff Sedimentation Cones, SAN, BrandTech Scientific, Inc.
Supplier: Brandtech
388000
89140-950EA 89.47 USD
89140-950
BRAND® Imhoff Sedimentation Cones, SAN, BrandTech Scientific, Inc.
Funnels Sedimentation Cones
For measurement of settleable/sedimenting solids in suspension.
  • Glass-clear SAN plastic for easy reading
  • Screw cap for draining
  • Withstands temperatures to 85 °C
These BRAND® Imhoff Sedimentation Cones are made of glass-clear styrene acrylonitrile copolymer with molded graduations for easy measurement of settled solids. Finely graduated up to 100 ml, with graduations every 50 ml up to 1000 ml. Screw cap assists drainage.
Certifications: Meets DIN 12672.