VWR® Economy Digital Thermometer
Supplier: VWR International
89140-510
89140-510EA 54.46 USD
89140-510
VWR® Economy Digital Thermometer
Thermometers Digital Thermometers
Economy digital thermometer with probe and used in ambient temperature.
  • High or low alarm throughput
  • With minimum and maximum memory
  • Available in °C and °F