VWR® Tall Microscope Slide Boxes for 100 Slides
Supplier: VWR International
HS15979A
89140-280CS 2020.51 USD
89140-280
VWR® Tall Microscope Slide Boxes for 100 Slides
Microscope Slide Boxes
Tall slide boxes are designed for storage and transport of non-standard microscope slides (3x2").
  • Grooved slots
  • Soft cork lining
  • Sturdy ABS construction for rugged use
Grooved slots hold individual slides in place. The box lid provides the needed clearance for the taller height of non-standard slides. Soft cork lining insures that your slides will not slip. Sturdy ABS construction for rugged use.