VWR® Slide Box
Supplier: VWR International
89140-278
89140-278CS 4157.29 USD
89140-278
VWR® Slide Box
Microscope Slide Boxes
Durable polycarbonate construction and rugged stainless steel provide safe storage and transport of 3x1" and 75x25 mm slides.
  • Polycarbonate and stainless steel construction
  • Durable design
  • Temperature-resistant foam lining
  • Stackable
Grooved slide slots keep slides separated, preventing contact. Temperature-resistant foam lining protects slides in the ultra low temerature range, down to -80°C. For easy slide retrieval, identification sheet, lining, and grooved slots are numbered.