VWR® Cargo Manual Pipette Rack
Supplier: VWR International
89140-276
89140-276CS 939.01 USD
89140-276
VWR® Cargo Manual Pipette Rack
Pipette Stands
The Cargo Manual pipette Rack clicks together horizontally with side connections, and is then securely locked in place with top connections.
  • Attaches to metal surfaces with magnets
  • Angled shelves safely store pipettes in a range of sizes
Magnets on both sides of the units connect securely and safely to metal surfaces such as file cabinets, fume hoods, refrigerators, and freezers. Angled shelves store 0.1–50 mL pipettes within arm’s reach for use. A protective lid encloses the unit to prevent dust build up. The lid also provides an additional shelf for extra space when connected horizontally. Varying shades of blue bring diversity to the lab.