D-(+)-Biotin

Supplier: Biotium

90072
89139-648EA 57.83 USD
89139-648
D-(+)-Biotin
D(+)-Biotin
Formula: C₁₀H₁₆N₂O₃S
Melting Pt: 232…233 °C
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005541
CAS Number: 58-85-5
EINECS: 200-399-3
Merck Index: 13,01231

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR