5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE) ≥98% (by TLC) amine-reactive fluorescent dye

Supplier: Biotium

Synonyms: TAMRA SE, 5 isomer, 5-TAMRA, SE, 5-TAMRA-N-succinimidyl ester, 5-Carboxytetramethylrhodamine N-succinimidyl ester

90034
89139-576EA 235.47 USD
89139-576
5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE) ≥98% (by TLC) amine-reactive fluorescent dye
5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE)

Full name: 5-Carboxytetramethylrhodamine succinimidyl ester (single isomer). Amine-reactive form of carboxytetramethylrhodamine (TAMRA) single isomer.


  • λEx/λEm (MeOH) = 540/565 nm
  • Dark red solid soluble in DMSO or DMF
  • Store at -20°C and protect from light

Formula: C₂₉H₂₅N₃O₇
MW: 527.52 g/mol
Storage Temperature: Freezer
CAS Number: 150810-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR