5(6)-TAMRA SE
Supplier: Biotium
Carboxytetramethylrhodamine is one of the most commonly used red fluorescent dye. The succinimidyl ester reacts readily with primary or secondary amines under mild conditions.