Sulforhodamine G fluorescent dye, for histology

Supplier: Biotium

80102
89139-506EA 165.87 USD
89139-506
Sulforhodamine G fluorescent dye, for histology
Sulforhodamine G

Similar to other sulforhodamine dyes, the highly water-soluble sulforhodamine G can also be used as a polar tracer for the studies of cell morphology and neuronal cell-cell communication.


  • λEx/λ Em = 529/548 nm
  • Orange red solid soluble in water
  • Store at room temperature

Storage Temperature: Ambient
MDL Number: MFCD00213704
CAS Number: 5873-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR