3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] membrane stain, for histology

Supplier: Biotium

Warning

70009
89139-376EA 275.23 USD
89139-376
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] membrane stain, for histology
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide]

DiOC6(3) (3,3′-Dihexyloxacarbocyanine,iodide) is a green fluorescent membrane dye that has been used to stain the ER in both live and fixed cells. The dye has also been used to study structural interactions and dynamics of the ER in neurons and yeast. In addition, the dye is widely used as a membrane potential dye.


  • λEx/λEm (MeOH) = 484/501 nm
  • Orange solid soluble in DMSO or DMF
  • Store at 4°C and protect from light, especially in solution

Formula: C₂₉H₃₇IN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00011954
CAS Number: 53213-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR