NBD C6-Sphingomyelin membrane stain, for histology

Supplier: Biotium

60031
89139-336EA 409.93 USD
89139-336
NBD C6-Sphingomyelin membrane stain, for histology
NBD C6-Sphingomyelin

NBD C6-sphingomyelin (6-((N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)amino)hexanoyl)sphingosyl phosphocholine) has been used for following sphingolipid metabolism in cells.


  • λEx/λEm (MeOH) = 466/536 nm
  • Dark solid soluble in chloroform or DMSO
  • Store at -20°C and protect from light

Formula: C₃₅H₆₁N₆O₉P
Storage Temperature: Freezer
CAS Number: 94885-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR