Pluronic® F-127 20% in DMSO

Supplier: Biotium

59004
89139-288EA 32.78 USD
89139-288
Pluronic® F-127 20% in DMSO
Poloxamer 407 (Pluronic® F-127)

Ready-to-use solution of Pluronic® F-127, a nonionic detergent useful for solubilizing hydrophobic dye molecules such as cell-permeable AM esters of Fura-2, Fluo-3, Indo-1, or Rhod-2 calcium indicators to facilitate cell loading.


We also offer Pluronic® F-127 solid and Pluronic® F-127, 10% in H2O.


Pluronic F-127 is a registered trade mark of BASF.

Formula: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
MDL Number: MFCD00082049
CAS Number: 9003-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR