Pluronic® F-127
Supplier: Biotium
Pluronic® F-127 is a nonionic detergent useful for solubilizing hydrophobic dye molecules such as cell-permeable AM esters of Fura-2, Fluo-3, Indo-1, or Rhod-2 calcium indicators to facilitate cell loading. Pluronic® F-127 is usually dissolved in water or DMSO with gentle heating.
- White solid soluble in water and DMSO
We also offer ready-to-use solutions of Pluronic® F-127, 20% in DMSO and Pluronic® F-127, 10% in H2O.
Pluronic F-127 is a registered trade mark of BASF.
Formula:
HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH Storage Temperature: Ambient |
MDL Number:
MFCD00082049 CAS Number: 9003-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...