5(6)-Carboxy-2',7'-dichlorofluorescein (CDCF) ≥98% (by TLC) fluorescent pH indicator

Supplier: Biotium

Synonyms: Carboxy-DCF, 5(6)-CDCF

51015
89139-252EA 145.7 USD
89139-252
5(6)-Carboxy-2',7'-dichlorofluorescein (CDCF) ≥98% (by TLC) fluorescent pH indicator
5(6)-Carboxy-2',7'-dichlorofluorescein (CDCF)

5-(and-6)-carboxy-2′,7′-dichlorofluorescein is similar to carboxyfluorescein (51013), but has lower pKa. It is useful for detecting pH in more acidic environment.


  • pKa = 4.8
  • λExλEm = 504/529 nm
  • ε= 90,000
  • Cell permeability: Membrane impermeant
  • Cellular localization:Fluid phase tracer
  • Colors: Green
  • Orange solid soluble in DMSO and pH>6 buffers
  • Store at room temperature and protect from light, especially in solution


Also see the membrane-permeant compound dichlorofluorescein diacetate (51016).

Formula: C₂₁H₁₀Cl₂O₇
Storage Temperature: Ambient
MDL Number: MFCD00037452
CAS Number: 111843-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR