BCECF AM ester 1 mg/mL in anhydrous DMSO ≥95% (by TLC) fluorescent pH indicator

Supplier: Biotium

Synonyms: 2′,7′-Bis(2-carboxyethyl)-5(6′)-carboxyfluorescein acetoxymethyl ester, Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-2',7'-dipropanoic acid, BCECF AM

51009
89139-238EA 157.08 USD
89139-238
BCECF AM ester 1 mg/mL in anhydrous DMSO ≥95% (by TLC) fluorescent pH indicator
BCECF AM ester

BCECF, AM ester is membrane-permeant and thus can be loaded into cells via incubation. This product is a mixture of three molecular species, all of which are readily hydrolyzed by endogenous esterases into a single BCECF free acid form once they are in the cells.


  • Cell permeability: Membrane permeant
  • Absorption/Emission λabs <300nm (before hydrolysis)
  • Colors: Green
  • λabs\λem = 503/528 nm (pH 9, after hydrolysis)
  • Store at -20°C and protect from light and moisture

Formula: C₄₂H₄₀O₂₁
Storage Temperature: Freezer
MDL Number: MFCD00036969
CAS Number: 117464-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR