Rhod-2 AM (Bromide) fluorescent indicator, for calcium determination

Supplier: Biotium

50023 50024
89139-194EA 342.21 USD
89139-194 89139-196
Rhod-2 AM (Bromide) fluorescent indicator, for calcium determination
Rhod-2 AM (Bromide)

Membrane-permeant form of Rhod-2 that can be loaded into cells via incubation. Because of the relatively low water solubility of the AM ester, the mild detergent Pluronic F-127 is often used as a dispersing agent to facilitate dye loading. Rhod-2-AM itself does not bind Ca2+ , but it is readily hydrolyzed to Rhod-2 by endogenous esterases once it enters cells.


  • Absorption/Emission 556nm/576nm (low or high [Ca2+] )
  • Extinction Coefficient 80,000 M-1 cm-1 (556 nm)
  • Cell permeability: Membrane permeant
  • Red solid soluble in DMSO
  • Store at -20°C and protect from light, especially in solution

Formula: C₅₂H₅₉BrN₄O₁₉
Storage Temperature: Freezer
CAS Number: 145037-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR