Ethidium bromide 1% in aqueous solution

Supplier: Biotium

Danger

40042
89139-116EA 106.66 USD
89139-116
Ethidium bromide 1% in aqueous solution
Ethidium bromide
Formula: C₂₁H₂₀BrN₃
MDL Number: MFCD00011724
CAS Number: 1239-45-8
EINECS: 214-984-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR