7-Aminoactinomycin D (7-AAD)

Supplier: Biotium

40037
89139-106EA 208.7 USD
89139-106
7-Aminoactinomycin D (7-AAD)
7-Aminoactinomycin D (7-AAD)
Formula: C₆₂H₈₇N₁₃O₁₆
MDL Number: MFCD00077295
CAS Number: 7240-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b98a49d6dcbe8c124f36e266a132074a5b18b08a&vid=b98a49d6dcbe8c124f36e266a132074a5b18b08a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR