5-Bromo-2'-deoxyuridine (broxuridine) 95% (by HPLC), off-white solid

Supplier: Biotium

Synonyms: 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU

40024
89139-084EA 74.9 USD
89139-084
5-Bromo-2'-deoxyuridine (broxuridine) 95% (by HPLC), off-white solid
5-Bromo-2'-deoxyuridine (Broxuridine)

BrdU (5-Bromo-2′-deoxyuridine) can be incorporated into DNA during cell division and subsequently detected by a BrdU antibody. The probe can be used to study cell-cycle kinetics.

Formula: C₉H₁₁BrN₂O₅
MW: 307.10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006529
CAS Number: 59-14-3
EINECS: 200-415-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR