Ponceau S 0.1 % (w/v) in 5% acetic acid staining solution

Supplier: Biotium

22001
89138-848EA 130.23 USD
89138-848
Ponceau S 0.1 % (w/v) in 5% acetic acid staining solution
Ponceau S

Ponceau S staining solution is used for the detection of proteins on PVDF and nitrocellulose membranes with a limit of detection of about 250 ng of transferred protein.


  • Rapidly stains proteins on membranes pink or light red
  • For easy visual inspection of protein transfer
  • Reversible and compatible with subsequent Western blotting

Formula: C₂₂H₁₂N₄Na₄O₁₃S₄
MDL Number: MFCD00003892
CAS Number: 6226-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR