5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Supplier: Biotium
10001
10001-2
89138-180EA
84.69
USD
89138-180
89138-184
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Formula:
C₈H₄BrClNNa₂O₄P |
MDL Number:
MFCD00036757 CAS Number: 102185-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...