French Square Bottles, Narrow Mouth, Glass, Clear, C&G Containers
Supplier: C&G Containers
LFF004400000 LFF004900100 LFF008900000 LFF016900100 LFF032900000 LFF002308400
71000-762CS 486.83 USD
71000-762 71000-764 71000-766 71000-768 71000-772 71000-774 71000-776 89137-862
French Square Bottles, Narrow Mouth, Glass, Clear, C&G Containers
Bottles Glass Bottles
These glass jars are used primarily for the collection of sediment sampling.
Pre-cleaned and certified products contain the certificate of analysis and cleaning protocol form in the product case.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for pre-cleaned and certified products. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (Nitric acid wash), B1 (Deionized water wash), C1 (Hydrochloric acid wash), and L3 (no wash).
They can accommodate a mix of both water and sediments containing granules of rock, oil, or petroleum distillates, and are ideally suited for collection of water samples for mulitple analyte testing. Please refer to the applicable certificate of analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity 'contain' volumes may vary.