Lab-Tips® Small Knitted Polyester Swab with Flexible Tip
Supplier: Berkshire
LTP70F.20
89137-163CS 281.86 USD
89137-163
Lab-Tips® Small Knitted Polyester Swab with Flexible Tip
Swabs Swabs for Cleaning
The LTP70F Swab is an excellent swab for cleaning grooved, slotted, and recessed areas.
  • Cleanroom laundered, double layer, 100% polyester knit swab
  • Critically low particles, fibers, ions, and extractables
  • High abrasion resistance
  • Chemically compatible with IPA, acetone, and other solvents
  • Soft and non-abrasive with no contaminating adhesives
  • Good sorbency
  • Free from Silicone, amides, and DOP
  • Autoclavable
The LTP70F Swab features a cleanroom laundered, knitted polyester head that is extremely clean and durable. A compact handle and internal flexible paddle provide maximum dexterity in confined areas.
Swabs are laundered and packaged in a Class 10 (ISO Class 4) cleanroom.