Accu-Safe Enclosed Chamber Bottle Thermometers, Thermco
Supplier: Thermco
ACCULF010S ACCF0101S ACCR0201S ACCBBS ACCI0301S ACCDB301 ACCDB40 ACCI300S ACCI310S ACCR20001S ACCOV700S ACCOV070S ACCOV080S ACCBB210S ACCF100S ACCAM0401S
89137-076EA 102.36 USD
89137-076 89137-078 89137-080 89137-082 89137-084 89137-086 89137-088 10145-490 10145-492 10145-494 10145-496 10145-498 10145-500 10145-502 10145-550 10145-556
Accu-Safe Enclosed Chamber Bottle Thermometers, Thermco
Thermometers Analog Thermometers
These spirit filled, non-mercury thermometers are housed in unbreakable plastic bottles filled with a non-toxic insulating liquid, used to ensure accurate temperature readings without being affected by the opening and closing of a chamber door.
Thermometers are fluoropolymer coated for additional protection. All thermometers have thoroughly annealed glass and engraved stems for lasting accuracy in 0.1, 0.5 and 1 °C divisions. Magnets are included to attach thermometers to a steel door or wall.
Ordering information: All thermometers have their own unique serial number and are supplied with a NIST traceable statement.