Activate™ Bleach Dilution System, Deardorff Fitzsimmons
Supplier: Deardorff Fitzsimmons Corp
77062
89136-482EA 177.83 USD
89136-482
Activate™ Bleach Dilution System, Deardorff Fitzsimmons
Cleaning Agents Disinfectants
The innovative Activate™ Bleach Dilution System dispenses the 10% bleach solution widely recommended by CDC and other nationally recognized authorities
This two-bottle fixed-ratio trigger sprayer automatically mixes and dilutes 5.25% bleach concentrate with tap water to make a 5000 ppm bleach disinfectant solution for use on hard, non-porous surfaces. The sprayer keeps the bleach separate from the water until you spray, keeping the 10% solution fresh and effective. Refill the water cartridge with tap water and lock in the disposable bleach cartridges and system is ready for use.
Its fast contact times, sporicidal action and innovative dispenser make it the most practical way to use bleach in the healthcare and laboratory environment.
Ordering information: Includes Six, 11 oz. Bleach Cartridges, Two 5000 ppm spayer heads, and Two 11 oz. Water Bottles.