BRAND PCR Microtubes with Attached Caps, BrandTech®
Supplier: Brandtech
781300
89135-430PK 232.45 USD
89135-430
BRAND PCR Microtubes with Attached Caps, BrandTech®
Tubes PCR Tubes
Unique cap design closes and opens easily without tools, yet ensures a tight fit to reduce sample evaporation.
  • Flat caps allow simple labeling
  • DNase-, RNase- and DNA-free, autoclavable 121 °C (20 min)
This 0.2 ml PCR tubes with attached caps are compatible with leading thermal cyclers with heated lids.