KleenGuard™ V80 Revolution™ Protective Goggles, Kimberly-Clark Professional
Supplier: Ansell Healthcare
14399
89135-145EA 26.47 USD
89135-145 10748-014
KleenGuard™ V80 Revolution™ Protective Goggles, Kimberly-Clark Professional
Goggles
When you need splash protection, anti-fog lenses, and a sleek look all rolled into one, trust KleenGuard™ Revolution™ OTG Safety Goggles.
  • Universal size (one size fits most)
  • Meets ANSI Z87.1+ standard for impact protection with a D3 rating for droplet and splash protection
  • Polycarbonate lenses provide 99.9% UVA/UVB/UVC protection
  • Indirect ventilation protects from splashes and provides increased airflow to minimize condensation and fogging
  • The lens is hardcoated for scratch protection and longer life
  • Fits over most prescription eyeglasses and features adjustable headband buckles for a snug fit
KleenGuard™ Revolution™ OTG Safety Goggles feature a soft, pliable frame gasket for enhanced fit and comfort, and cross-vent technology to keep air flowing and moisture out to prevent fogging. The clear lenses allow maximum light transmission and feature an anti-fog coating. They are made of lightweight polycarbonate material that provides 99.9% UVA/UVB/UVC protection, and are hardcoated for scratch resistance and longer life.
Certifications: These safety goggles meet the ANSI Z87.1+ standards for impact, with a D3 rating for droplet and splash protection, and are TAA compliant.