Exhalation Valve Assembly System Component, 3M™
Supplier: 3M
7000052738
89135-102CS 468.78 USD
89135-102
Exhalation Valve Assembly System Component, 3M™
Respirators Respiratory Systems
Exhalation valve assembly includes mounting seat and 3M™ Cool Flow™ Valve for use with 3M™ Ultimate FX Full Facepiece Reusable Respirators.
It is used for facility sanitation, food processing and food safety.