Gentamicin sulfate in solution for tissue culture

Supplier: IBI Scientific

Synonyms: Gentamicin sulfate salt, NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

IB02030
89135-046EA 77.67 USD
89135-046
Gentamicin sulfate in solution for tissue culture
Gentamicin sulphate

Gentamycin Sulfate is a broad spectrum antibiotic that is a more effective in-vitro bacterial inhibitor than combined streptomycin-penicillin


Refrigerated shipping and storage is required for this product.


Gentamycin is non-toxic to tissue culture monolayers, and does not inhibit virus replication. It inhibits the growth of a wide variety of gram-positive and gram-negative microorganisms, including strains resistant to tetracycline, chloramphenicol, kanamycin, and colistin; particularly strains of Pseudomonoas, Proteus, Staphylococcus, and Streptococcus. Gentamycin Sulfate inhibits bacterial protein biosynthesis by binding to the 30S subunit of the ribosome. Antibiotic inhibition/sensitivity assay: PASS.

MDL Number: MFCD00270181
CAS Number: 1405-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR