E.Z.N.A. PX Blood RNA Kit, Omega Bio-tek
Supplier: OMEGA BIO-TEK
R1057-00
89134-792KT
61.7
USD
89134-792
E.Z.N.A. PX Blood RNA Kit, Omega Bio-tek
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
Omega Bio-tek’s E.Z.N.A.® PX Blood RNA Kit is designed for isolation of total RNA from blood samples stored in special preserved reagents and Paxgene tubes
The procedure completely removes contaminants and enzyme inhibitors making RNA isolation fast, convenient, and reliable. RNA purified using the E.Z.N.A.® PX RNA Kit method is ready for applications such as RT-PCR.