Professional Buckets, Vileda Professional-FHP
Supplier: Vileda Professional - FHP
131586
89134-307CS 704.68 USD
89134-307
Professional Buckets, Vileda Professional-FHP
Buckets
Fits all popular cleanroom cart systems.
  • Available in blue or red for color coding
  • Ideal for 10" and 14" Roll-O-Matic Mops
  • Fits popular cleanroom cart systems
  • Buckets are autoclavable
  • Casters sold separately
Ideal for 25.4cm (10") and 35.6cm (14") Roll-O-Matic® mops and MicroPad wet and dry 45.7cm (18") pads. Available in blue or red for color coding. Buckets are completely autoclavable. Casters are available separately.