Catheter Locking Solution, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
HGS-10
89134-258EA 58.24 USD
89134-258
Catheter Locking Solution, SAI Infusion Technologies
Animal Supplies Animal Infusion Supplies
The antithrombotic characteristics of Heparin and the viscous nature of glycerol or dextrose are combined in a solution to promote and extend the patency of vascular catheters
  • Used in both large and small species
  • High osmolarity of dextrose prevents bacterial colonization
  • High viscosity reduces washout from the catheter tip
  • Effective physical barrier against clot formation
These solutions are proven performers in catheterized animal studies.
Ordering information: Minimum order of 5 bottles applies, also available in pre-filled syringes.